Order Entry
Poland
ContactUsLinkComponent
Anti-Sulfapyridine Sheep Polyclonal Antibody
Anti-Sulfapyridine Sheep Polyclonal Antibody
Catalog # USBIS8016-01
Supplier:  US Biological
Anti-Sulfapyridine Sheep Polyclonal Antibody
Catalog # USBIS8016-01
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Sulfapyridine
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Sheep
  • Form:
    liquid
  • Storage buffer:
    20mM PBS, 0,09% sodium azide, pH 7,2.
  • Storage temperature:
    Cool
  • Concentration:
    ~~1 mg/ml (UV abs)
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: ELISA: 10 ?g/ml

Type: Primary
Antigen: Sulfapyridine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: