Anti-STAT 5a Rabbit Polyclonal Antibody
50 µg
Catalog # USBIS7972-10R
Supplier: US Biological
Jump to:
Product Details & Documents
Recommended dilution: Western Blot:1:500-1:1000; Immunochemistry:1:50-1:100
Type: Primary
Antigen: STAT 5a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
Specifications
- Size:50 µg
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by immunoaffinity chromatography
- Antibody type:Primary
- Form:liquid
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Shipping temperature:Ice
- Cat. No.:USBIS7972-10R
- Reactivity:Human|Rat|Mouse
- Conjugation:Unconjugated
- Antigen symbol:STAT 5a
- Storage buffer:PBS(without Mg2+ and Ca2+), pH 7,4, 150mMsodium chloride, 0,02% sodium azide, 50% glycerol.



