Anti-Sentrin 2 Rabbit Polyclonal Antibody
Catalog # USBIS0750-12
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Sentrin 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,4, and 1% BSA with 0,1% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: Sentrin 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human