Anti-Semaphorin 5A Goat Polyclonal Antibody
Catalog # USBIS0668-57G
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Semaphorin 5A
- Antigen symbol:SEMA5A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:128000; Western Blot: 0. 03?g/ml; Immunochemistry: 4-6?g/ml
Type: Primary
Antigen: Semaphorin 5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Rat