Anti-Semaphorin 5A Sheep Polyclonal Antibody
Catalog # USBIS0668-57E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Semaphorin 5A
- Antigen symbol:SEMA5A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1 ?g/ml; ELISA: 0.5-1 ?g/ml
Type: Primary
Antigen: Semaphorin 5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat