Order Entry
Poland
ContactUsLinkComponent
Anti-Semaphorin 5A Mouse Polyclonal Antibody
Anti-Semaphorin 5A Mouse Polyclonal Antibody
  USBIS0668-57C
Anti-Semaphorin 5A Mouse Polyclonal Antibody
  USBIS0668-57C

 

  • Antibody type:
    Primary
  • Antigen name:
    Semaphorin 5A
  • Antigen symbol:
    SEMA5A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    Serum
  • Form:
    liquid
  • Storage buffer:
    50% glycerol
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

 

 

Type: Primary
Antigen: Semaphorin 5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human