Anti-Rhox-13 Reproductive Homeobox on X Chromosome 13, Reproductive Homeobox Rabbit Polyclonal Antibody
Catalog # USBIR1998-92
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Rhox-13 Reproductive Homeobox on X Chromosome 13, Reproductive Homeobox
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,05% BSA, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 2-4?g/ml
Type: Primary
Antigen: Rhox-13 Reproductive Homeobox on X Chromosome 13, Reproductive Homeobox
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse