Order Entry
Poland
ContactUsLinkComponent
Anti-Protocadherin beta-12 Mouse Monoclonal Antibody [clone: 9G428]
Anti-Protocadherin beta-12 Mouse Monoclonal Antibody [clone: 9G428]
Catalog # USBIP9119-40
Supplier:  US Biological
Anti-Protocadherin beta-12 Mouse Monoclonal Antibody [clone: 9G428]
Catalog # USBIP9119-40
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Protocadherin beta-12
  • Clonality:
    Monoclonal
  • Clone:
    9G428
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    lyophilized powder
  • Storage buffer:
    PBS, pH 7,2.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Protocadherin beta-12
Clonality: monoclonal
Clone: 9G428
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human