Anti-Protein Kinase A, alpha/beta, Catalytic Subunit Rabbit Polyclonal Antibody
USBIP9102-91J
- Antibody type:Primary
- Antigen name:Protein Kinase A, alpha/beta, Catalytic Subunit
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Modification:phospho-Thr197
- Storage buffer:Dulbecco’s PBS (without Mg2+ and Ca2+), pH 7,3, 1mg/ml BSA (IgG, protease free), 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:10 Pieces
- Pk:10
Recommended dilution: Western Blot: 0.1-0.75 ?g/ml
Type: Primary
Antigen: Protein Kinase A, alpha/beta, Catalytic Subunit
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human