Anti-Progesterone Mouse Monoclonal Antibody [clone: 10F316]
numer katalogowy USBIP9006-01E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Progesterone
- Clonality:Monoclonal
- Clone:10F316
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Tested applications:Radio Immunoassay
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Progesterone
Clonality: monoclonal
Clone: 10F316
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: