Anti-Progesterone Mouse Monoclonal Antibody [clone: 9L576]
Catalog # USBIP9006-01D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Progesterone
- Clonality:Monoclonal
- Clone:9L576
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Form:liquid
- Storage buffer:PBS, pH7.15, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~4 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Progesterone
Clonality: monoclonal
Clone: 9L576
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: