Anti-PI4K2A Rabbit Polyclonal Antibody
Catalog # USBIP4186-27P
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:phosphatidylinositol 4-kinase type 2 alpha
- Antigen symbol:PI4K2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Amino acid number:1 to 30
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:10-1:50
Type: Primary
Antigen: PI4K2A (phosphatidylinositol 4-kinase type 2 alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human