Anti-Peroxidase Anti-Peroxidase Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 5H17]
USBIP3350-31
- Antibody type:Tag
- Antigen name:Peroxidase Anti-Peroxidase
- Clonality:Monoclonal
- Clone:5H17
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- Isotype:IgG
- Reactivity:
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2 or .
- Storage temperature:Cool
- Concentration:~5 mg/ml
- Shipping temperature:Ice
- Purification:purified by delipidation, salt fractionation and ion exchange chromatography
- Size:5 mg
- Pk:5 mg
Type: Tag
Antigen: Peroxidase Anti-Peroxidase
Clonality: monoclonal
Clone: 5H17
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: