Order Entry
Poland
ContactUsLinkComponent
Anti-Papilloma Virus Type 18, E7 Mouse Monoclonal Antibody [clone: 9B108]
Anti-Papilloma Virus Type 18, E7 Mouse Monoclonal Antibody [clone: 9B108]
Catalog # USBIP3105-25C
Supplier:  US Biological
Anti-Papilloma Virus Type 18, E7 Mouse Monoclonal Antibody [clone: 9B108]
Catalog # USBIP3105-25C
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Papilloma Virus Type 18, E7
  • Clonality:
    Monoclonal
  • Clone:
    9B108
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~5,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    1 mg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Papilloma Virus Type 18, E7
Clonality: Monoclonal
Clone: 9B108
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human