Anti-Neurofilament M 145-160kD Mouse Monoclonal Antibody [clone: 6H9]
Catalog # USBIN2160-01R
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Neurofilament M 145-160kD
- Clonality:Monoclonal
- Clone:6H9
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Rat
- Western blot:Yes
- Format:Tissue culture supernatant
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide.
- Molecular weight:145 - 160 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Immunofluorescence: 1:100; Western Blot: 1:1000
Type: Primary
Antigen: Neurofilament M 145-160kD
Clonality: monoclonal
Clone: 6H9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat