Anti-Myoglobin, Human Mouse Monoclonal Antibody [clone: 7E121]
USBIM9800-21C
- Antibody type:Primary
- Antigen name:Myoglobin
- Clonality:Monoclonal
- Clone:7E121
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,3, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~2/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: Myoglobin, Human
Clonality: Monoclonal
Clone: 7E121
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human