Order Entry
Poland
ContactUsLinkComponent
Anti-MSH6, exon 10 Goat Polyclonal Antibody
Anti-MSH6, exon 10 Goat Polyclonal Antibody
Catalog # USBIM4692-36G
Supplier:  US Biological
Anti-MSH6, exon 10 Goat Polyclonal Antibody
Catalog # USBIM4692-36G
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MSH6, exon 10
  • Antigen symbol:
    MSH6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Goat
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    Tris-citrate/phosphate and 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Immunoprecipitation: 1-4?g/mg; Western Blot: 1:1000-1:10000

Type: Primary
Antigen: MSH6, exon 10
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human