Anti-MAP3K1 Rabbit Polyclonal Antibody
USBIM2867-01D
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase kinase kinase 1, E3 ubiquitin Protein ligase
- Antigen symbol:MAP3K1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:25mMsodium borate, 100mM boric acid, pH 8.4, 75mM sodium chloride, 5mM EDTA.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: MAP3K1 (mitogen-activated protein kinase kinase kinase 1, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human