Anti-LMX 1.2 Mouse Polyclonal Antibody
USBIL2200-02
- Antibody type:Primary
- Antigen symbol:LMX1.2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Format:Serum
- Form:liquid
- Storage buffer:50% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: LMX 1.2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human