Anti-LEP Rabbit Polyclonal Antibody
Catalog # USBIL1669-01S
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:leptin
- Antigen symbol:LEP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide before the addition of glycerol to 40%.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:250 µg
- Pk:250 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:2000
Type: Primary
Antigen: LEP (leptin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human