Order Entry
Poland
ContactUsLinkComponent
Anti-Keratin, pan 4, 5, 6, 8, 10, 13, 18 Mouse Monoclonal Antibody [clone: C-11]
Anti-Keratin, pan 4, 5, 6, 8, 10, 13, 18 Mouse Monoclonal Antibody [clone: C-11]
Catalog # USBIK0199-89A
Supplier:  US Biological
Anti-Keratin, pan 4, 5, 6, 8, 10, 13, 18 Mouse Monoclonal Antibody [clone: C-11]
Catalog # USBIK0199-89A
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Keratin, pan 4, 5, 6, 8, 10, 13, 18
  • Clonality:
    Monoclonal
  • Clone:
    C-11
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Donkey,
    Frog,
    Human,
    Rat
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 15mM sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Keratin, pan 4, 5, 6, 8, 10, 13, 18
Clonality: monoclonal
Clone: C-11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Burro, Frog, Human, Rat