Order Entry
Poland
ContactUsLinkComponent
Anti-Interleukin 18 Mouse Monoclonal Antibody [clone: 3H2454]
Anti-Interleukin 18 Mouse Monoclonal Antibody [clone: 3H2454]
Catalog # USBII8439-95E
Supplier:  US Biological
Anti-Interleukin 18 Mouse Monoclonal Antibody [clone: 3H2454]
Catalog # USBII8439-95E
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 18
  • Antigen symbol:
    IL18
  • Clonality:
    Monoclonal
  • Clone:
    3H2454
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Pig
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    500 µg
  • Pk:
    500 µG

Specifications

About this item

Recommended dilution: ELISA: 5 ?g/ml

Type: Primary
Antigen: Interleukin 18
Clonality: monoclonal
Clone: 3H2454
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pig