Anti-Interleukin 1 Receptor Type I Hamster Monoclonal Antibody [clone: JAMA-147]
Catalog # USBII7663-25D1A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 1 Receptor 1
- Antigen symbol:IL1R1
- Clonality:Monoclonal
- Clone:JAMA-147
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Hamster
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:25-1:100;
Type: Primary
Antigen: Interleukin 1 Receptor Type I
Clonality: monoclonal
Clone: JAMA-147
Conjugation: unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse