Anti-Integrin, beta1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 7H127]
Catalog # USBII7661-39A5
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Integrin beta 1
- Antigen symbol:ITGB1
- Clonality:Monoclonal
- Clone:7H127
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:50mMsodium Phosphate pH 7,5, 100mM Potassium chloride, 150mM NaCl, 5% glycerol, 0,2% BSA, 0,04% NaN3.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:120 Tests
- Pk:120 Tests
Specifications
About this item
Type: Primary
Antigen: Integrin, beta1
Clonality: monoclonal
Clone: 7H127
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human