Order Entry
Poland
ContactUsLinkComponent
Anti-Integrin, beta1 Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 6A267]
Anti-Integrin, beta1 Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 6A267]
Catalog # USBII7661-38R4
Supplier:  US Biological
Anti-Integrin, beta1 Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 6A267]
Catalog # USBII7661-38R4
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Integrin beta 1
  • Antigen symbol:
    ITGB1
  • Clonality:
    Monoclonal
  • Clone:
    6A267
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Hamster
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Form:
    lyophilized powder
  • Storage buffer:
    PBS, pH 7,4, 1% BSA, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Recommended dilution: Flow Cytometry:1:5;
Do not freeze!

Type: Primary
Antigen: Integrin, beta1
Clonality: monoclonal
Clone: 6A267
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse, Rat