Order Entry
Poland
ContactUsLinkComponent
Anti-Immunoglobulin-Like Transcript 7 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11D30]
Anti-Immunoglobulin-Like Transcript 7 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11D30]
Catalog # USBII7502-76
Supplier:  US Biological
Anti-Immunoglobulin-Like Transcript 7 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11D30]
Catalog # USBII7502-76
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Immunoglobulin-like transcript 7
  • Antigen symbol:
    ILT7
  • Clonality:
    Monoclonal
  • Clone:
    11D30
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    PBS,
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Do not freeze!

Type: Primary
Antigen: Immunoglobulin-Like Transcript 7
Clonality: monoclonal
Clone: 11D30
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human