Anti-Hydroxysteroid Dehydrogenase, 11-beta, Type I Rabbit Polyclonal Antibody
Catalog # USBIH9117-40A
Dostawca: US Biological
Dane techniczne
- Antibody type:Primary
- Antigen symbol:Hydroxysteroid Dehydrogenase, 11-beta, Type I
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:78 to 92
- Storage buffer:TBS, pH 7,4, 0,5mg/ml BSA, 0,02% sodium azide, 50% glycerol.
- Sequence:CLELGAASAHYIAGT
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Dane techniczne
Informacje o tym produkcie
Recommended dilution: Western Blot: 1:200; Immunochemistry: 1:200
Type: Primary
Antigen: Hydroxysteroid Dehydrogenase, 11-beta, Type I
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat