Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody [clone: LN3]
USBIH6100-42Q
- Antibody type:Primary
- Antigen name:HLA Class 2 Antigen DR
- Clonality:Monoclonal
- Clone:LN3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: HLA Class 2 Antigen DR
Clonality: monoclonal
Clone: LN3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human