Order Entry
Poland
ContactUsLinkComponent
Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: 5F62]
Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: 5F62]
Catalog # USBIH6099-95M
Supplier:  US Biological
Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: 5F62]
Catalog # USBIH6099-95M
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HLA Class 2 Antigen DR
  • Clonality:
    Monoclonal
  • Clone:
    5F62
  • Conjugation:
    PE (Phycoerythrin)/Cy5®
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    50 Tests
  • Pk:
    50 Tests

Specifications

About this item

Type: Primary
Antigen: HLA Class 2 Antigen DR
Clonality: monoclonal
Clone: 5F62
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human