Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 3H2224]
Catalog # USBIH6099-95G
Dostawca: US Biological
Dane techniczne
- Antibody type:Primary
- Antigen name:HLA Class 2 Antigen DR
- Clonality:Monoclonal
- Clone:3H2224
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG3
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2, 1.0% BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~100Tests/1ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 Tests
- Pk:100 Tests
Dane techniczne
Informacje o tym produkcie
Do not freeze!
Type: Primary
Antigen: HLA Class 2 Antigen DR
Clonality: monoclonal
Clone: 3H2224
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human