Anti-HLA Class 1 Antigen G Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MEM-G/11]
Catalog # USBIH6098-72T1
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:HLA Class 1 Antigen G
- Clonality:Monoclonal
- Clone:MEM-G/11
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, 15mM sodium azide, pH 7,4
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Recommended dilution: Flow Cytometry:1:500
Type: Primary
Antigen: HLA Class 1 Antigen G
Clonality: monoclonal
Clone: MEM-G/11
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human