Order Entry
Poland
ContactUsLinkComponent
Anti-HLA-DR+DP Mouse Monoclonal Antibody [clone: 6D461]
Anti-HLA-DR+DP Mouse Monoclonal Antibody [clone: 6D461]
Catalog # USBIH6098-72D
Supplier:  US Biological
Anti-HLA-DR+DP Mouse Monoclonal Antibody [clone: 6D461]
Catalog # USBIH6098-72D
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HLA-DR DP
  • Clonality:
    Monoclonal
  • Clone:
    6D461
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 15mM sodium azide, before the addition of glycerol to 40%.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: HLA-DR+DP
Clonality: monoclonal
Clone: 6D461
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human