Anti-Fluorescein Isothiocyanate Rabbit polyclonal antibody
USBIF5250-11G
- Antibody type:Tag
- Antigen name:Fluorescein isothiocyanate
- Antigen symbol:FITC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:1 mg
- Pk:1 mg
Type: Tag
Antigen: Fluorescein Isothiocyanate
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: