Anti-FLT3 Rabbit Polyclonal Antibody (FITC (Fluorescein))
USBIF4600-07C-FITC
- Antibody type:Primary
- Antigen name:Fms-related tyrosine kinase 3
- Antigen symbol:FLT3
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein G chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: FLT3 (fms-related tyrosine kinase 3)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human