Anti-F9 Mouse monoclonal antibody [clone: 13F42-F6]
USBIF0017-05A
- Antibody type:Primary
- Antigen name:coagulation factor IX
- Antigen symbol:F9
- Clonality:Monoclonal
- Clone:13F42-F6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:0,05M sodium phosphate, 0,1M sodium chloride, 1mM EDTA, pH 6.6.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:250 µg
- Pk:250 µG
Recommended dilution: Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: F9 (coagulation factor IX)
Clonality: monoclonal
Clone: 13F42-F6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human