Order Entry
Poland
ContactUsLinkComponent
Anti-Endoplasmic Reticulum Protein 57 Mouse monoclonal antibody [clone: 4E69]
Anti-Endoplasmic Reticulum Protein 57 Mouse monoclonal antibody [clone: 4E69]
Catalog # USBIE2291-75-200
Supplier:  US Biological
Anti-Endoplasmic Reticulum Protein 57 Mouse monoclonal antibody [clone: 4E69]
Catalog # USBIE2291-75-200
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Endoplasmic Reticulum Protein 57
  • Clonality:
    Monoclonal
  • Clone:
    4E69
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Hamster,
    Pig,
    Monkey,
    Human,
    Dog
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 25mM sodium chloride, 5mM sodium phosphate, 0,05mM PMSF, 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    200 µg
  • Pk:
    200 µG

Specifications

About this item

Recommended dilution: Western Blot: 0.5 ?g/ml; Immunoprecipitation: 10 ?g/ml

Type: Primary
Antigen: Endoplasmic Reticulum Protein 57
Clonality: monoclonal
Clone: 4E+69
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Hamster, Human, Monkey, Pig