Order Entry
Poland
ContactUsLinkComponent
 
Anti-DHEA 15 Rabbit polyclonal antibody
  USBID3226-25
Anti-DHEA 15 Rabbit polyclonal antibody
  USBID3226-25

 

  • Antibody type:
    Primary
  • Antigen name:
    Dehydroepiandrosterone 15
  • Antigen symbol:
    DHEA15
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Format:
    Serum
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    1 Kit
  • Pk:
    1 KIT

 

 

Recommended dilution: 1:100000

Type: Primary
Antigen: DHEA 15
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: