Anti-C6 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
USBIC7850-35G-APC
- Antibody type:Primary
- Antigen name:complement component 6
- Antigen symbol:C6
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Amino acid number:37 to 66
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Flow Cytometry: 1:10-1:50; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: C6 (complement component 6)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human