Anti-Complement C3a Receptor Mouse Monoclonal Antibody [clone: 10B380]
Catalog # USBIC7850-13T
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Complement C3a Receptor
- Clonality:Monoclonal
- Clone:10B380
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:10-1:20;
Type: Primary
Antigen: Complement C3a Receptor
Clonality: Monoclonal
Clone: 10B380
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human