Anti-Chlamydia trachomatis MOMP Goat Polyclonal Antibody (FITC (Fluorescein))
Catalog # USBIC4250-55
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Chlamydia trachomatis MOMP
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Form:liquid
- Storage buffer:PBS, pH 7,2, 1% BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~5 mg/ml
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Primary
Antigen: Chlamydia trachomatis MOMP
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: