Order Entry
Poland
ContactUsLinkComponent
 
Anti-THBD Mouse Monoclonal Antibody [clone: QBEND/40]
Catalog # USBIC2548-75E
Supplier:  US Biological
Anti-THBD Mouse Monoclonal Antibody [clone: QBEND/40]
Catalog # USBIC2548-75E
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Thrombomodulin
  • Antigen symbol:
    THBD
  • Clonality:
    Monoclonal
  • Clone:
    QBEND/40
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Format:
    Supernatant
  • Form:
    liquid
  • Storage buffer:
    0,09% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Recommended dilution: Immunochemistry: 1:10-1:100

Type: Primary
Antigen: THBD (thrombomodulin)
Clonality: Monoclonal
Clone: QBEND/40
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human