Anti-ITGAE (Integrin alpha E, CD103) Hamster Monoclonal Antibody (Biotin) [clone: 2E7]
USBIC2445-65E
- Antibody type:Primary
- Antigen name:Integrin Alpha E
- Antigen symbol:ITGAE
- Clonality:Monoclonal
- Clone:2E7
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Hamster
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:500 µg
- Pk:500 µG
Type: Primary
Antigen: ITGAE (Integrin alpha E, CD103)
Clonality: Monoclonal
Clone: 2E7
Conjugation: Biotin
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse