Anti-CD70 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H173]
USBIC2425-01P
- Antibody type:Primary
- Antigen name:CD70 molecule
- Antigen symbol:CD70
- Clonality:Monoclonal
- Clone:7H173
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,5, 0,2% BSA, 0,04% sodium azide,15% glycerol.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Size:120 Tests
- Pk:120 Tests
Recommended dilution: Flow Cytometry: 1:50
Do not freeze!
Type: Primary
Antigen: CD70 (CD70 molecule)
Clonality: Monoclonal
Clone: 7H173
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human