Anti-MS4A1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5F23]
USBIC2270-08A
- Antibody type:Primary
- Antigen name:Membrane-spanning 4-Domains, Subfamily A, Member 1
- Antigen symbol:MS4A1
- Clonality:Monoclonal
- Clone:5F23
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG3
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 Tests
- Pk:50 Tests
Type: Primary
Antigen: MS4A1 (membrane-spanning 4-domains, subfamily A, member 1)
Clonality: Monoclonal
Clone: 5F23
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human