Anti-CD5 Mouse Monoclonal Antibody [clone: MEM-32]
Catalog # USBIC2256-25M
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:CD5 molecule
- Antigen symbol:CD5
- Clonality:Monoclonal
- Clone:MEM-32
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 15mM sodium azide, pH 7,4.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 20 ?g/ml
Type: Primary
Antigen: CD5 (CD5 molecule)
Clonality: Monoclonal
Clone: MEM-32
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human