Anti-CD3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 7H222]
Catalog # USBIC2254-01E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:CD3
- Clonality:Monoclonal
- Clone:7H222
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 15% glycerol, 0,2% BSA, 0,05% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:120 Tests
- Pk:120 Tests
Specifications
About this item
Do not freeze!
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 7H222
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human