Anti-CD3, epsilon Hamster Monoclonal Antibody [clone: 145-2C11]
Catalog # USBIC2253-38A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:CD3 epsilon
- Clonality:Monoclonal
- Clone:145-2C11
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Hamster
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:250 µg
- Pk:250 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:25-1:200
Type: Primary
Antigen: CD3, epsilon
Clonality: Monoclonal
Clone: 145-2C11
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse