Anti-BAI3 Mouse monoclonal antibody unconjugated [clone: 8H174]
Catalog # USBIB2704-12C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:brain-specific angiogenesis inhibitor 3
- Antigen symbol:BAI3
- Clonality:Monoclonal
- Clone:8H174
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:lyophilized powder
- Amino acid number:25 to 880
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 0.5-1.0 ?g/ml; Immunochemistry: 25 ?g/ml
Type: Primary
Antigen: BAI3 (brain-specific angiogenesis inhibitor 3)
Clonality: monoclonal
Clone: 8H174
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human