Anti-BAI3 Rabbit polyclonal antibody unconjugated
USBIB2704-10D
- Antibody type:Primary
- Antigen name:brain-specific angiogenesis inhibitor 3
- Antigen symbol:BAI3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Form:liquid
- Amino acid number:1506 to 1522
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: ELISA: 1:1000; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: BAI3 (brain-specific angiogenesis inhibitor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human