Anti-Bone Morphogenetic Protein 11 Mouse monoclonal antibody unconjugated [clone: 9G320]
USBIB2553-38
- Antibody type:Primary
- Antigen name:Bone Morphogenetic Protein 11
- Antigen symbol:BMP11
- Clonality:Monoclonal
- Clone:9G320
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Form:lyophilized powder
- Amino acid number:310 to 407
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Bone Morphogenetic Protein 11
Clonality: monoclonal
Clone: 9G320
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human